Learn More
Biomatik Corporation Human Kinectin 1 (KTN1) ELISA Kit
Supplier:  Biomatik Corporation EKU09127
Encompass_Preferred
Human Kinectin 1 (KTN1) ELISA Kit for detecting KTN1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 30pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q86UP2. Gene ID: 3895. Target Synonyms: CG1; KNT; Kinesin Receptor; CG-1 antigen; Kinesin receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8564