Learn More
Biomatik Corporation Human Kinesin Family, Member 1A (KIF1A) ELISA Kit
Supplier:  Biomatik Corporation EKU05497
Encompass_Preferred
Human Kinesin Family, Member 1A (KIF1A) ELISA Kit for detecting KIF1A in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q12756. Gene ID: 547. Target Synonyms: ATSV; UNC104; C2orf20; Axonal Transport Of Synaptic Vesicles; Microtubule-based motor KIF1A; Unc-104- and KIF1A-related protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6904