Learn More
Biomatik Corporation Human Kinesin Light Chain 1 (KLC1) ELISA Kit
Supplier:  Biomatik Corporation EKU05494
Encompass_Preferred
Human Kinesin Light Chain 1 (KLC1) ELISA Kit for detecting KLC1 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.106ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q07866. Gene ID: 3831. Target Synonyms: KLC; KNS2; KNS2A; hKLC1S; hKLC1N; hKLC1P; hKLC1G; hKLC1R; hKLC1J; hKLC1B; Kinesin 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8902