Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Kininogen ELISA Kit

Catalog No. p-7219481
Encompass_Preferred
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests

ELISA

Human Kininogen quantitates human Kininogen in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Kininogen.

This gene uses alternative splicing to generate two different proteins- high molecular weight kininogen (HMWK) and low molecular weight kininogen (LMWK). HMWK is essential for blood coagulation and assembly of the kallikrein-kinin system. Also, bradykinin, a peptide causing numerous physiological effects, is released from HMWK. In contrast to HMWK, LMWK is not involved in blood coagulation. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Alpha-1-MAP; alpha-2-thiol proteinase inhibitor; BDK; BK; Bradykinin; fitzgerald factor; high molecular weight kininogen; H-kininigen; HMWK; Ile-Ser-Bradykinin; Kallidin I; Kallidin II; kininogen; kininogen 1; kininogen 1-like 1; kininogen-1; Kininogen-1 heavy chain; Kininogen-1 light chain; Kng; KNG1; Kng1l1; L-kininogen; low molecular weight (LMW) T-kininogen II; Low molecular weight growth-promoting factor; Lysyl-bradykinin; Major acute phase protein; major acute phase protein (alpha-1-MAP); Thiostatin; T-kinin; T-kininogen 2; T-kininogen 2 heavy chain; T-kininogen 2 light chain; T-kininogen II; T-kininogen II heavy chain; T-kininogen II light chain; Williams-Fitzgerald-Flaujeac factor
Assay Range 24 to 6,000 ng/mL
Assay Sensitivity 24 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3827
Gene Symbol KNG1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Kininogen 1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.