Learn More
Sino Biological Human OX-40L / TNFSF4 / CD252 Protein (ECD, Fc Tag) 500ug
Supplier:  Sino Biological 13127H01H500
Encompass_Preferred
A DNA sequence encoding the human TNFSF4 (Gln51-Leu183) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-222-3646