Learn More
Sino Biological Human KIR2DL1 MAb 25TS (Sino Biological)
Supplier:  Sino Biological 13145R124C25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human KIR2DL1 / CD158a (rh KIR2DL1 / CD158a; Catalog#13145-H08H; NP_055033.2; Met1-His245) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Catalog No. 50-237-2797