Learn More
Sino Biological Recombinant Anti-KIR2DL1 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 13145R124P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human KIR2DL1 / CD158a (rh KIR2DL1 / CD158a; Catalog#13145-H08H; NP_055033.2; Met1-His245) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-255-4301