Learn More
Sino Biological Anti-KIR2DL1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13145T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human KIR2DL1 / CD158a (rh KIR2DL1 / CD158a; Catalog#13145-H08H; NP_055033.2; Met1-His245). KIR2DL1 / CD158a specific IgG was purified by Human KIR2DL1 / CD158a affinity chromatography.
Catalog No. 50-255-4298