Learn More
Sino Biological Human KIR2DL3 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 12828RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human KIR2DL3 (rh KIR2DL3; Catalog#12828-H08H; AAX23102.1; Met 1-His 245). KIR2DL3 specific IgG was purified by Human KIR2DL3 affinity chromatography.
Catalog No. 50-237-2095