Learn More
Sino Biological Recombinant Human KIR2DL3 Protein (ECD, hFc Tag)
Supplier:  Sino Biological 12828H02H100
Encompass_Preferred
A DNA sequence encoding the human KIR2DL3 (AAX23102.1) (Met1-His245) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-4343