Learn More
Sino Biological Anti-KIR2DL4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13052RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human KIR2DL4 / CD158D (rh KIR2DL4 / CD158D; Catalog#13052-HCCS; ADY38409.1; Met1-His242). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4337