Learn More
Sino Biological Anti-KIRREL2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 15674T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human KIRREL2 / NEPH3 (rh KIRREL2 / NEPH3; Catalog#15674-H08H; NP_115499.4; Met1-Arg503). KIRREL2 / NEPH3 specific IgG was purified by Human KIRREL2 / NEPH3 affinity chromatography.
Catalog No. 50-241-2314