Learn More
Sino Biological Anti-c-Kit Antibody (PE), Mouse Monoclonal
Catalog No. 502412166
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human c-Kit (rh c-Kit; Catalog#11996-H08H; P10721-2; Met1-Thr516) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-2166 Supplier Sino Biological Supplier No. 11996MM34P100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human c-Kit (rh c-Kit; Catalog#11996-H08H; P10721-2; Met1-Thr516) and conjugated with PE under optimum conditions, the unreacted PE was removed.