Learn More
Sino Biological Anti-c-Kit Antibody (HRP), Mouse Monoclonal
Supplier:  Sino Biological 11996MM14H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human KIT / c-KIT / CD117 (rh KIT / c-KIT / CD117; Catalog#11996-H08H; P10721-2; Met1-Thr516). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-241-2170