Learn More
Sino Biological Human KIT MAb 20ul (Sino Biological)
Supplier:  Sino Biological 11996R35120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human c-Kit / CD117 (rh c-Kit / CD117; Catalog#11996-H08H; P10721-2; Met1-Thr516).
Catalog No. 50-237-2127