Learn More
Sino Biological Recombinant Anti-c-Kit Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11996R01820
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human KIT / c-KIT / CD117 (rh KIT / c-KIT / CD117; Catalog#11996-H08H; P10721-2; Met1-Thr516).
Catalog No. 50-241-1482