Learn More
Sino Biological Anti-c-Kit Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 11996MM41A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human c-Kit (rh c-Kit; Catalog#11996-H08H; P10721-2; Met1-Thr516) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-2221