Learn More
Sino Biological Recombinant Anti-c-Kit Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 11996R007A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human c-Kit / CD117 (rh c-Kit / CD117; Catalog#11996-H08H; P10721-2; Met1-Thr516) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-2228