Learn More
Sino Biological Human KIT MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11996R007100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human KIT / c-KIT / CD117 (rh KIT / c-KIT / CD117; Catalog#11996-H08H; P10721-2; Met1-Thr516).
Catalog No. 50-237-1458