Learn More
Sino Biological Anti-c-Kit Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11996RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human c-Kit / CD117 / KIT (rh c-Kit / CD117 / KIT; Catalog#11996-H08H; P10721-2; Met 1-Thr 516). c-Kit / CD117 / KIT specific IgG was purified by Human c-Kit / CD117 / KIT affinity chromatography.
Catalog No. 50-241-1486