Learn More
Sino Biological Human KITLG MAb 20ul (Sino Biological)
Supplier:  Sino Biological 10451R00420
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human SCF / C-kit ligand (rh SCF / C-kit ligand; Catalog#10451-H08B; P21583-1; Met1-Ala189).
Catalog No. 50-237-1195