Learn More
Sino Biological Anti-SCF Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 207249T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human SCF fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-2281