Learn More
BIOHIPPO Human KL-6 Microsample
SDP
Catalog No. 5000314147
Encompass
Human KL-6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of KL-6 in serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 U/mL with a sensitivity of 14 25 U/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human KL-6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human KL-6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human KL-6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14147 Supplier BIOHIPPO Supplier No. BHE15212418
Human KL-6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of KL-6 in serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 U/mL with a sensitivity of 14 25 U/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human KL-6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human KL-6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human KL-6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color