Learn More
Sino Biological Anti-KLF17 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 105186T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human KLF17, and purified by antigen affinity chromatography.
Catalog No. 50-241-0303