Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human KLF4-TAT Recombinant Protein, PeproTech®

Catalog No. 11008100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
5 μg
5 x 100 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11008100UG 100 μg
110-08-25UG 25 μg
11-008-500 5 x 100 μg
110-08-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 11008100UG Supplier Gibco™ Supplier No. 11008100UG
  • $690.00
    $517.65 / Each of 1

    Save $172.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

110-08-1MG will be provided as 10 x 100 μg (110-08-100UG). 110-08-250UG will be provided as multiple 25 μg (110-08-25UG) and 100 μg (110-08-100UG) vials. 110-08-500UG will be provided as 5 x 100 μg (110-08-100UG). Recombinant Human KLF4-TAT is a 483 amino acid protein, including a 13-residue C-terminal TAT peptide, with a calculated molecular weight of 51.7 kDa. PeproTech's Recombinant Human KLF4-TAT is a mixture of the expected sequence beginning at Met1 and a truncated isoform beginning at Tyr54. Due to post-translational modifications, SDS-PAGE gel shows bands at approximately 72 and 66kDa, under reduced conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.

KLF4 is a member of the Kruppel-like factor (KLF) family of zinc finger transcription factors. Members of this family share 3 contiguous C2H2-type zinc fingers at the carboxyl terminus that comprise the DNA-binding domain. KLF4 is highly expressed in skin and gut epithelial tissues, but is also found in various other cells and tissues, including vascular endothelial cells, lymphocytes, lung, and testis. It is an important regulator of the cell cycle, transcription, and cell differentiation. Together with Sox2, Oct4, and cMyc, KLF4 can induce the reprogramming of primary human fibroblasts to a pluripotent state. KLF4 and other transcription factors can be introduced into cells by DNA transfection, viral infection, or microinjection. Protein transduction using TAT fusion proteins represents an alternative methodology for introducing transcription factors into primary, as well as transformed, cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43474
For Use With (Application) Control
Formulation Protein with no preservative
Gene ID (Entrez) 9314
Molecular Weight (g/mol) 51.7 kDa
Name Human KLF4-TAT
Purification Method Purified
Quantity 100 μg
Source HEK293 cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias KLF4
Common Name KLF4-TAT
Gene Symbol KLF4
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag TAT-fusion
Sequence MAVSDALLPS FSTFASGPAG REKTLRQAGA PNNRWREELS HMKRLPPVLP GRPYDLAAAT VATDLESGGA GAACGGSNLA PLPRRETEEF NDLLDLDFIL SNSLTHPPES VAATVSSSAS ASSSSSPSSS GPASAPSTCS FTYPIRAGND PGVAPGGTGG GLLYGRESAP PPTAPFNLAD INDVSPSGGF VAELLRPELD PVYIPPQQPQ PPGGGLMGKF VLKASLSAPG SEYGSPSVIS VSKGSPDGSH PVVVAPYNGG PPRTCPKIKQ EAVSSCTHLG AGPPLSNGHR PAAHDFPLGR QLPSRTTPTL GLEEVLSSRD CHPALPLPPG FHPHPGPNYP SFLPDQMQPQ VPPLHYQELM PPGSCMPEEP KPKRGRRSWP RKRTATHTCD YAGCGKTYTK SSHLKAHLRT HTGEKPYHCD WDGCGWKFAR SDELTRHYRK HTGHRPFQCQ KCDRAFSRSD HLALHMKRHF GGYGRKKRRQ RRR
Content And Storage -20°C
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 90% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.