Learn More
Raybiotech Inc Human Kallikrein 10 ELISA Kit
Supplier:  Raybiotech Inc ELHKLK101
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 2 ng/mL. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human KLK10. Other species not determined.
Catalog No. 50-194-9477