Learn More
Sino Biological Anti-Kallikrein 11 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10767RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human KLK11 / Kallikrein 11 (rh KLK-11; Catalog#10767-H08H; NP_006844.1; Met 1-Asn 250). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1356