Learn More
Sino Biological Recombinant Anti-KLK3/PSA Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10771R103100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human KLK3 / Kallikrein 3 (rh KLK3 / Kallikrein 3; Catalog#10771-H08H; P07288; Met1-Pro261).
Catalog No. 50-240-9383