Learn More
Sino Biological Anti-KLK3/PSA Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10771MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human KLK3 / Kallikrein 3 (rh KLK3 / Kallikrein 3; Catalog#10771-H08H; P07288; Met1-Pro261). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-0565