Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KLK3 native Protein from human seminal plasma

Catalog No. 89950093 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-950-093 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-950-093 Supplier Abnova™ Supplier No. P4872
Add to Cart
Add to Cart

Used for SDS-PAGE, WB-Ti

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. Its protein product is a protease present in seminal plasma. It is thought to function normally in the liquefaction of seminal coagulum, presumably by hydrolysis of the high molecular mass seminal vesicle protein. Serum level of this protein, called PSA in the clinical setting, is useful in the diagnosis and monitoring of prostatic carcinoma. Alternate splicing of this gene generates several transcript variants encoding different isoforms. [provided by RefSeq]

Specifications

For Use With (Application) SDS-PAGE, Western Blot
Formulation No additive
Gene ID (Entrez) 354
Molecular Weight (g/mol) 30kDa
Name KLK3 (Human) Native Protein
Preparation Method Native protein purified from human seminal plasma
Purification Method Gel filtration/Ion exchange chromatography purification
Quantity 100 μg
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with deionized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias APS/KLK2A1/PSA/hK3
Common Name KLK3
Gene Symbol KLK3
Species Human
Protein Tag None
Expression System Native protein purified from human seminal plasma
Form Lyophilized
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.