Learn More
Sino Biological Human KLK3/PSA Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785482
Encompass_Preferred
The Human KLK3 / Kallikrein 3 ELISA Pair Set is for the quantitative determination of Human KLK3 / Kallikrein 3.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human KLK3 / Kallikrein 3 coated on a 96-well plate. Standards and samples are added to the wells, and any Human KLK3 / Kallikrein 3 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human KLK3 / Kallikrein 3 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human KLK3 / Kallikrein 3 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85482 Supplier Sino Biological Supplier No. SEK107715
The Human KLK3 / Kallikrein 3 ELISA Pair Set is for the quantitative determination of Human KLK3 / Kallikrein 3.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human KLK3 / Kallikrein 3 coated on a 96-well plate. Standards and samples are added to the wells, and any Human KLK3 / Kallikrein 3 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human KLK3 / Kallikrein 3 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human KLK3 / Kallikrein 3 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.