Learn More
Sino Biological Anti-KLK4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 205415T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human KLK4 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-240-7705