Learn More
Sino Biological Recombinant Human Kallikrein 7/KLK7 Protein (His Tag)
Supplier:  Sino Biological 10416H08H50
Encompass_Preferred
A DNA sequence encoding the human KLK7 (NP_005037.1) extracellular domain (Met 1-Arg 253) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-224-6936