Learn More
American Research Products Inc Human Klotho ELISA Kit
Supplier:  American Research Products Inc CSBE13235H
Encompass
Human Klotho ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29870