Learn More
Biomatik Corporation Human Klotho (KL) CLIA Kit
Supplier:  Biomatik Corporation EKU09978
Encompass_Preferred
Human Klotho (KL) CLIA Kit for detecting KL in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 41.2-30,000pg/ml. Sensitivity: 16.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9UEF7. Gene ID: 9365. Target Synonyms: KLa; Klotho Alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8389