Learn More
Sino Biological Recombinant Human KLRB1 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 18474H01H100
Encompass_Preferred
A DNA sequence encoding the Human KLRB1 (Q12918) (Gln67-Ser225) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-241-2567