Learn More
Sino Biological Anti-NKG2A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13905T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human NKG2A / NKG2 / CD159A / KLRC1 (rh NKG2A / NKG2 / CD159A / KLRC1; Catalog#13905-H07H; NP_002250.1; Arg100-Leu233). NKG2A / NKG2 / CD159A / KLRC1 specific IgG was purified by Human NKG2A / NKG2 / CD159A / KLRC1 affinity chromatography.
Catalog No. 50-255-4168