Learn More
Sino Biological Recombinant Human NKp80/KLRF1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10984H07H100
Encompass_Preferred
A DNA sequence encoding the Human KLRF1 (NP_057607.1) (Leu60-Tyr231) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-4994