Learn More
Sino Biological Human KLRK1 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 10575H04B1
Encompass_Preferred
A DNA sequence encoding the Human KLRK1 (P26718-1) (Phe78-Val216) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-237-1730