Learn More
Sino Biological Anti-Kininogen 1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10529MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human KNG1 / Kininogen 1 (rh KNG1 / Kininogen 1; Catalog#10529-H08H; NP_001095886.1; Gln19-Ser644). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3615