Learn More
Sino Biological Anti-Kininogen 1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 201463T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human Kininogen 1/KNG1 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-2262