Learn More
ACROBiosystems Protein, Human KRAS, G12C, His Tag, 50 μg
Catalog No. 502253103
Encompass_Preferred
This human KRAS (2-185, G12C) protein is a small GTPase transductor that binds GDP/GTP and possesses intrinsic GTPase activity. Expressed from E. coli cells, this active enzyme protein carries a polyhistidine tag at the N-terminus and is lyophilized with trehalose as a protectant.
  • Expressed from E. coli, containing amino acids Thr 2 to Cys 185
  • Verified purity of >95% by SDS-PAGE and >90% by SEC-HPLC
  • Calculated molecular weight of 23.0 kDa, migrating as 23-25 kDa under reducing conditions
  • Lyophilized formulation from 0.22 μm filtered solution in 20 mM Tris, 500 mM NaCl, pH 7.5
  • Stable for 12 months at -20°C to -70°C in lyophilized state
  • Active enzyme validated by GTPase-Glo assay
This item is not returnable. View return policy
Catalog No. 50-225-3103 Supplier ACROBiosystems Supplier No. KRSH51H350UG
This human KRAS (2-185, G12C) protein is a small GTPase transductor that binds GDP/GTP and possesses intrinsic GTPase activity. Expressed from E. coli cells, this active enzyme protein carries a polyhistidine tag at the N-terminus and is lyophilized with trehalose as a protectant.
  • Expressed from E. coli, containing amino acids Thr 2 to Cys 185
  • Verified purity of >95% by SDS-PAGE and >90% by SEC-HPLC
  • Calculated molecular weight of 23.0 kDa, migrating as 23-25 kDa under reducing conditions
  • Lyophilized formulation from 0.22 μm filtered solution in 20 mM Tris, 500 mM NaCl, pH 7.5
  • Stable for 12 months at -20°C to -70°C in lyophilized state
  • Active enzyme validated by GTPase-Glo assay