Learn More
ACROBiosystems Human KRAS 50ug
Catalog No. 502534534
Encompass_Preferred
Human KRAS (G12V) Protein His Tag (KRS-H5143) is expressed from E. coli cells. It contains AA Thr 2 - Cys 185 (Accession # P01116-2 (G12V)).
Catalog No. 50-253-4534 Supplier ACROBiosystems Supplier No. KRSH514350UG
Human KRAS (G12V) Protein His Tag (KRS-H5143) is expressed from E. coli cells. It contains AA Thr 2 - Cys 185 (Accession # P01116-2 (G12V)).