Learn More
AMID BIOSCIENCES Amid Biosciences, Human KRAS G13D Protein, Recombinant, Biotinylated, 0.1 mg
SDP
Catalog No. NC4043453
Encompass_Preferred
Human KRAS G13D is supplied as a recombinant biotinylated protein in a 0.1 mg format for oncology research and RAS pathway assay development. Applications include SPR, BLI, ELISA, Pull-down, AlphaScreen, MST, protein-protein interaction studies, drug discovery workflows. Technical specifications are included below.- Quantity - 0.1 mg- Gene - KRAS- Mutation / Variant - G13D- Organism - Human- Tag - N-terminal AviTag, N-terminal 8XHis- Expression System - E. coli- Purity - greater than 90% by Coomassie staining- UniProt ID - P01116-2- Amino Acid Range - 1-185- Buffer - 20 mM HEPES, pH 7.5, 100 mM NaCl, 1 mM MgCl2, 1 mM 2-mercapthoethanol, 10% glycerol- Endotoxin Level - less than 1 EU/mg- Storage - -80 C- Biotinylated - Yes, N-terminusThis product is for research use only.
Catalog No. NC4043453 Supplier AMID BIOSCIENCES Supplier No. KRAS13DB3011
Human KRAS G13D is supplied as a recombinant biotinylated protein in a 0.1 mg format for oncology research and RAS pathway assay development. Applications include SPR, BLI, ELISA, Pull-down, AlphaScreen, MST, protein-protein interaction studies, drug discovery workflows. Technical specifications are included below.- Quantity - 0.1 mg- Gene - KRAS- Mutation / Variant - G13D- Organism - Human- Tag - N-terminal AviTag, N-terminal 8XHis- Expression System - E. coli- Purity - greater than 90% by Coomassie staining- UniProt ID - P01116-2- Amino Acid Range - 1-185- Buffer - 20 mM HEPES, pH 7.5, 100 mM NaCl, 1 mM MgCl2, 1 mM 2-mercapthoethanol, 10% glycerol- Endotoxin Level - less than 1 EU/mg- Storage - -80 C- Biotinylated - Yes, N-terminusThis product is for research use only.