Learn More
Sino Biological Recombinant Human KRAS Protein (His Tag) 500
Supplier:  Sino Biological 12259H07E6500
Encompass_Preferred
A DNA sequence encoding the human KRAS (P01116-2) (Thr2-Cys185, with natural variant Gly 12 Asp) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-224-6996