Learn More
Sino Biological Human KRAS / K-Ras (G12D & Q61H) Protein (His Tag) 1
Supplier:  Sino Biological 12259H07E11
Encompass_Preferred
A DNA sequence encoding the human KRAS (P01116-2) (Thr2-Cys185, with natural variant Gly 12 Asp and Gln 61 His) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-224-7088