Learn More
Sino Biological Recombinant Human KRAS Protein (His & AVI Tag), Biotinylated
Supplier:  Sino Biological 12259H56EB20
Encompass_Preferred
A DNA sequence encoding the human KRAS (P01116-2) (Thr2-Cys185(G12D)) was expressed with a N-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-1568