Learn More
BIORTUS USA INC Human KRas4B Protein, biotinylated/GDP_loaded, 100UG
Catalog No. NC2953333
Encompass_Preferred
Gene Name: KBTBD6.Uniprot: Q86V97.Construct: GG-KBTBD6(Q2-Q674 end). Expression Host: Insect cell. Purity: 85%.
Catalog No. NC2953333 Supplier BIORTUS USA INC Supplier No. BP192030DB
Gene Name: KBTBD6.Uniprot: Q86V97.Construct: GG-KBTBD6(Q2-Q674 end). Expression Host: Insect cell. Purity: 85%.