Learn More
BIORTUS USA INC Human KRAS4B Protein, biotinylated/GDP_loaded, 100UG
Catalog No. NC2953309
Encompass_Preferred
Gene Name: KRAS. Uniprot: P01116. Construct: biotinylated-Avi-KRas4B(T2-M188 end) (G12D),GDP loaded. Expression Host: E. coli. Purity: 90%.
Catalog No. NC2953309 Supplier BIORTUS USA INC Supplier No. BP190730DB
Gene Name: KRAS. Uniprot: P01116. Construct: biotinylated-Avi-KRas4B(T2-M188 end) (G12D),GDP loaded. Expression Host: E. coli. Purity: 90%.