Learn More
BIORTUS USA INC Human KRas4B Protein, biotinylated, 100UG
Catalog No. NC2953322
Encompass_Preferred
Gene Name: KRAS. Uniprot: P01116. Construct: biotinylated-Avi-KRas4B(T2-M188 end), GMPPNP loaded. Expression Host: E. coli. Purity: 95%.
Catalog No. NC2953322 Supplier BIORTUS USA INC Supplier No. BP192030MB
Gene Name: KRAS. Uniprot: P01116. Construct: biotinylated-Avi-KRas4B(T2-M188 end), GMPPNP loaded. Expression Host: E. coli. Purity: 95%.